siRNA Details
| Gene Symbol: Azgp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012826.2 | NP_036958.2 | 4 | 768 | ||
| XM_006249003.2 | XP_006249065.1 | 6 | 949 | |||
| XM_006249004.2 | XP_006249066.1 | 6 | 933 | |||
| GenBank mRNA | BC105822.1 | - | 4 | 768 | 7371444 | |
| D21058.1 | - | 4 | 767 | |||
| FQ210978.1 | - | 787 | ||||
| FQ219509.1 | - | 4 | 759 | |||
| X75309.1 | - | 4 | 749 | |||

Azgp1
Genomic Map