siRNA Details
Gene Symbol: Abcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012833.2 | NP_036965.1 | 3 | 498 | ||
GenBank mRNA | D86086.1 | - | 3 | 320 | ||
L49379.1 | - | 3 | 379 | |||
X96393.1 | - | 3 | 334 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Abcc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012833.2 | NP_036965.1 | 3 | 498 | ||
GenBank mRNA | D86086.1 | - | 3 | 320 | ||
L49379.1 | - | 3 | 379 | |||
X96393.1 | - | 3 | 334 |