siRNA Details
| Gene Symbol: Comp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012834.1 | NP_036966.1 | 17 | 2039 | ||
| XM_006252822.2 | XP_006252884.1 | 14 | 2243 | |||
| GenBank mRNA | X72914.1 | - | 17 | 2039 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Comp| Gene Symbol: Comp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012834.1 | NP_036966.1 | 17 | 2039 | ||
| XM_006252822.2 | XP_006252884.1 | 14 | 2243 | |||
| GenBank mRNA | X72914.1 | - | 17 | 2039 | ||