siRNA Details
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012836.2 | NP_036968.1 | 4 | 1209 | ||
GenBank mRNA | AF284830.1 | - | 4 | 566 | ||
U62897.1 | - | 4 | 1209 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012836.2 | NP_036968.1 | 4 | 1209 | ||
GenBank mRNA | AF284830.1 | - | 4 | 566 | ||
U62897.1 | - | 4 | 1209 |