siRNA Details
| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X54737.1 | - | 3 | 379 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cstb| Gene Symbol: Cstb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | X54737.1 | - | 3 | 379 | ||