siRNA Details
| Gene Symbol: Cyct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012840.3 | NP_036972.1 | 369 | |||
| GenBank mRNA | M20623.1 | - | 1 | 199 | ||
| Gene Symbol: Pde11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127480.1 | NP_001120952.1 | 3 | 371 | ||
| GenBank mRNA | AB059361.1 | - | 3 | 371 | ||

Cyct
Genomic Map