siRNA Details
| Gene Symbol: Ephx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034090.3 | NP_001029262.1 | 4, 5 | 735 | ||
| NM_012844.3 | NP_036976.2 | 4, 5 | 764 | |||
| GenBank mRNA | BC061568.1 | - | 4, 5 | 695 | 6889368 | |
| M26125.1 | - | 740 | ||||

Ephx1
Genomic Map