siRNA Details
| Gene Symbol: Lamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012857.2 | NP_036989.1 | 2 | 214 | ||
| GenBank mRNA | M34959.1 | - | 2 | 214 | ||
| X14765.1 | - | 1 | 50 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lamp1| Gene Symbol: Lamp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012857.2 | NP_036989.1 | 2 | 214 | ||
| GenBank mRNA | M34959.1 | - | 2 | 214 | ||
| X14765.1 | - | 1 | 50 | |||