siRNA Details
| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012859.1 | NP_036991.1 | 9 | 2850 | ||
| XM_006228391.2 | XP_006228453.1 | 9 | 1983 | |||
| XM_006228393.2 | XP_006228455.1 | 9 | 2234 | |||
| XM_006228394.2 | XP_006228456.1 | 9 | 3561 | |||
| XM_008758895.1 | XP_008757117.1 | 9 | 1967 | |||
| XM_008758896.1 | XP_008757118.1 | 9 | 2127 | |||
| GenBank mRNA | U40001.1 | - | 9 | 2850 | ||
| X51415.1 | - | 8 | 2372 | |||

Lipe
Genomic Map