siRNA Details
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012861.1 | NP_036993.1 | 3 | 295 | ||
GenBank mRNA | M76704.1 | - | 2 | 240 | ||
S61804.1 | - | 3 | 295 | |||
X54862.1 | - | 3 | 321 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mgmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012861.1 | NP_036993.1 | 3 | 295 | ||
GenBank mRNA | M76704.1 | - | 2 | 240 | ||
S61804.1 | - | 3 | 295 | |||
X54862.1 | - | 3 | 321 |