siRNA Details
| Gene Symbol: Bhlha15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012863.1 | NP_036995.1 | 2 | 369 | ||
| XM_008768941.1 | XP_008767163.1 | 2 | 1030 | |||
| GenBank mRNA | BC061868.1 | - | 2 | 317 | 5598799 | |
| U58279.1 | - | 2 | 369 | |||

Bhlha15
Genomic Map