siRNA Details
Gene Symbol: Bhlha15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012863.1 | NP_036995.1 | 2 | 3415 | ||
GenBank mRNA | U58279.1 | - | 2 | 3415 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bhlha15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012863.1 | NP_036995.1 | 2 | 3415 | ||
GenBank mRNA | U58279.1 | - | 2 | 3415 |