siRNA Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012868.1 | NP_037000.1 | 1 | 195 | ||
| XM_006232045.2 | XP_006232107.1 | 1 | 779 | |||
| GenBank mRNA | L27339.1 | - | 1 | 195 | ||
| X75605.1 | - | 1 | 216 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npr3| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012868.1 | NP_037000.1 | 1 | 195 | ||
| XM_006232045.2 | XP_006232107.1 | 1 | 779 | |||
| GenBank mRNA | L27339.1 | - | 1 | 195 | ||
| X75605.1 | - | 1 | 216 | |||