siRNA Details
| Gene Symbol: Npr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012868.1 | NP_037000.1 | 3 | 933 | ||
| XM_006232045.2 | XP_006232107.1 | 3 | 1517 | |||
| GenBank mRNA | L27339.1 | - | 3 | 933 | ||
| X75605.1 | - | 3 | 954 | |||
| X78595.1 | - | 779 | ||||

Npr3
Genomic Map