siRNA Details
| Gene Symbol: Ros1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012874.1 | NP_037006.1 | 8 | 1173 | ||
| XM_008772862.1 | XP_008771084.1 | 8 | 1354 | |||
| XM_008772863.1 | XP_008771085.1 | 12 | 1082 | |||
| GenBank mRNA | M35104.1 | - | 8 | 1173 | ||
| M35105.1 | - | 8 | 1173 | |||
| M35106.1 | - | 8 | 1173 | |||

Ros1
Genomic Map