siRNA Details
| Gene Symbol: LOC103692785 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770106.1 | XP_008768328.1 | 1 | 206 | ||
| Gene Symbol: LOC103694169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771991.1 | XP_008770213.1 | 1 | 202 | ||
| Gene Symbol: Rpl39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012875.2 | NP_037007.1 | 172 | |||
| GenBank mRNA | BC058489.1 | - | 206 | 6887843 | ||
| X82551.1 | - | 172 | ||||

LOC103692785
Genomic Map