siRNA Details
| Gene Symbol: Sftpd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012878.2 | NP_037010.1 | 7 | 1161 | ||
| XM_008771065.1 | XP_008769287.1 | 7 | 1164 | |||
| GenBank mRNA | BC070507.1 | - | 8 | 1170 | 7098194 | |
| M81231.1 | - | 8 | 1168 | |||

Sftpd
Genomic Map