siRNA Details
| Gene Symbol: Spp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012881.2 | NP_037013.2 | 7 | 1249 | ||
| XM_008769996.1 | XP_008768218.1 | 7 | 1322 | |||
| GenBank mRNA | BC078874.1 | - | 7 | 1229 | 7110677 | |
| FQ213473.1 | - | 7 | 1238 | |||
| FQ222192.1 | - | 7 | 1265 | |||
| M14656.1 | - | 7 | 1221 | |||
| M99252.1 | - | 7 | 1257 | |||

Spp1
Genomic Map