siRNA Details
| Gene Symbol: Tmpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012887.1 | NP_037019.1 | 7 | 1163 | ||
| XM_008765219.1 | XP_008763441.1 | 5 | 986 | |||
| GenBank mRNA | U18314.1 | - | 7 | 1163 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmpo| Gene Symbol: Tmpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012887.1 | NP_037019.1 | 7 | 1163 | ||
| XM_008765219.1 | XP_008763441.1 | 5 | 986 | |||
| GenBank mRNA | U18314.1 | - | 7 | 1163 | ||