siRNA Details
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012888.1 | NP_037020.1 | 10 | 3116 | ||
GenBank mRNA | M34842.1 | - | 10 | 3116 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tshr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012888.1 | NP_037020.1 | 10 | 3116 | ||
GenBank mRNA | M34842.1 | - | 10 | 3116 |