siRNA Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012891.2 | NP_037023.1 | 4 | 285 | ||
| GenBank mRNA | BC087036.1 | - | 4 | 306 | 7193535 | |
| D30647.1 | - | 4 | 287 | |||
| FQ210754.1 | - | 4 | 334 | |||
| FQ219329.1 | - | 4 | 330 | |||

Acadvl
Genomic Map