siRNA Details
| Gene Symbol: Adk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012895.3 | NP_037027.2 | 3 | 171 | ||
| XM_006251631.1 | XP_006251693.1 | 3 | 285 | |||
| GenBank mRNA | BC081712.1 | - | 3 | 171 | 7193044 | |
| U90340.1 | - | 3 | 174 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adk| Gene Symbol: Adk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012895.3 | NP_037027.2 | 3 | 171 | ||
| XM_006251631.1 | XP_006251693.1 | 3 | 285 | |||
| GenBank mRNA | BC081712.1 | - | 3 | 171 | 7193044 | |
| U90340.1 | - | 3 | 174 | |||