siRNA Details
| Gene Symbol: Atf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012912.2 | NP_037044.1 | 4 | 585 | ||
| GenBank mRNA | BC078903.1 | - | 4 | 585 | 7100767 | |
| M63282.1 | - | 4 | 590 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atf3| Gene Symbol: Atf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012912.2 | NP_037044.1 | 4 | 585 | ||
| GenBank mRNA | BC078903.1 | - | 4 | 585 | 7100767 | |
| M63282.1 | - | 4 | 590 | |||