siRNA Details
| Gene Symbol: Atp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012914.1 | NP_037046.1 | 12, 13 | 1684 | ||
| XM_006246597.2 | XP_006246659.1 | 13, 14 | 2298 | |||
| GenBank mRNA | M30581.1 | - | 12, 13 | 1684 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp2a3| Gene Symbol: Atp2a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012914.1 | NP_037046.1 | 12, 13 | 1684 | ||
| XM_006246597.2 | XP_006246659.1 | 13, 14 | 2298 | |||
| GenBank mRNA | M30581.1 | - | 12, 13 | 1684 | ||