siRNA Details
| Gene Symbol: Alx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012921.1 | NP_037053.1 | 2 | 741 | ||
| XM_006241293.2 | XP_006241355.1 | 3 | 553 | |||
| GenBank mRNA | BC158591.1 | - | 2 | 517 | 9023971 | |
| L14018.1 | - | 2 | 741 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alx1| Gene Symbol: Alx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012921.1 | NP_037053.1 | 2 | 741 | ||
| XM_006241293.2 | XP_006241355.1 | 3 | 553 | |||
| GenBank mRNA | BC158591.1 | - | 2 | 517 | 9023971 | |
| L14018.1 | - | 2 | 741 | |||