siRNA Details
| Gene Symbol: Casp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012922.2 | NP_037054.1 | 3 | 135 | ||
| XM_006253130.2 | XP_006253192.1 | 3 | 179 | |||
| GenBank mRNA | BC081854.1 | - | 3 | 135 | 7104685 | |
| U49930.1 | - | 2 | 105 | |||
| U84410.1 | - | 2 | 109 | |||

Casp3
Genomic Map