siRNA Details
| Gene Symbol: Bcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012931.1 | NP_037063.1 | 1 | 294 | ||
| GenBank mRNA | D29766.1 | - | 1 | 294 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bcar1| Gene Symbol: Bcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012931.1 | NP_037063.1 | 1 | 294 | ||
| GenBank mRNA | D29766.1 | - | 1 | 294 | ||