siRNA Details
| Gene Symbol: Bcar1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012931.1 | NP_037063.1 | 2 | 700 | ||
| XM_006255628.2 | XP_006255690.1 | 2 | 544 | |||
| XM_006255629.2 | XP_006255691.1 | 2 | 2501 | |||
| GenBank mRNA | D29766.1 | - | 2 | 700 | ||
| FQ212404.1 | - | 2 | 502 | |||
| FQ221713.1 | - | 2 | 488 | |||

Bcar1
Genomic Map