siRNA Details
| Gene Symbol: Crmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012932.2 | NP_037064.1 | 4 | 598 | ||
| XM_006251084.2 | XP_006251146.1 | 4 | 832 | |||
| GenBank mRNA | FM031363.1 | - | 4 | 598 | ||
| U52095.1 | - | 4 | 392 | |||
| U52102.1 | - | 4 | 566 | |||

Crmp1
Genomic Map