siRNA Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012934.1 | NP_037066.1 | 4, 5 | 549 | ||
| XM_006254670.2 | XP_006254732.1 | 4, 5 | 1129 | |||
| XM_006254671.2 | XP_006254733.1 | 4, 5 | 760 | |||
| GenBank mRNA | AF389425.1 | - | 4, 5 | 549 | ||
| AF398465.1 | - | 4, 5 | 818 | |||
| U52104.1 | - | 4, 5 | 765 | |||

Dpysl3
Genomic Map