siRNA Details
| Gene Symbol: Crybb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012936.2 | NP_037068.2 | 5 | 587 | ||
| GenBank mRNA | AF286652.1 | - | 4 | 432 | ||
| AW919139.1 | - | 5 | 587 | |||
| BC126096.1 | - | 4 | 460 | 8364407 | ||
| X05900.1 | - | 4 | 293 | |||

Crybb1
Genomic Map