siRNA Details
| Gene Symbol: Ggh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012960.2 | NP_037092.1 | 2 | 306 | ||
| GenBank mRNA | BC087602.1 | - | 2 | 306 | 7312627 | |
| U38379.1 | - | 2 | 156 | |||
| Gene Symbol: LOC102549738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006237929.2 | XP_006237991.1 | 2 | 311 | ||

Ggh
Genomic Map