siRNA Details
| Gene Symbol: Ggh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012960.2 | NP_037092.1 | 5 | 627 | ||
| GenBank mRNA | BC087602.1 | - | 5 | 627 | 7312627 | |
| U38379.1 | - | 5 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ggh| Gene Symbol: Ggh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012960.2 | NP_037092.1 | 5 | 627 | ||
| GenBank mRNA | BC087602.1 | - | 5 | 627 | 7312627 | |
| U38379.1 | - | 5 | 477 | |||