siRNA Details
| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012964.2 | NP_037096.2 | 3, 4 | 260 | ||
| GenBank mRNA | AF133037.1 | - | 3, 4 | 260 | ||
| AF336825.1 | - | 3, 4 | 260 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hmmr| Gene Symbol: Hmmr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012964.2 | NP_037096.2 | 3, 4 | 260 | ||
| GenBank mRNA | AF133037.1 | - | 3, 4 | 260 | ||
| AF336825.1 | - | 3, 4 | 260 | |||