siRNA Details
| Gene Symbol: Msx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012982.3 | NP_037114.2 | 2 | 702 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Msx2| Gene Symbol: Msx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012982.3 | NP_037114.2 | 2 | 702 | ||