siRNA Details
| Gene Symbol: Nxph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012994.2 | NP_037126.1 | 3 | 567 | ||
| GenBank mRNA | BC105770.1 | - | 3 | 567 | 7930348 | |
| L27867.1 | - | 2 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nxph1| Gene Symbol: Nxph1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012994.2 | NP_037126.1 | 3 | 567 | ||
| GenBank mRNA | BC105770.1 | - | 3 | 567 | 7930348 | |
| L27867.1 | - | 2 | 473 | |||