siRNA Details
| Gene Symbol: Pax6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013001.2 | NP_037133.1 | 12 | 2002 | ||
| XM_006234633.2 | XP_006234695.1 | 14 | 2427 | |||
| XM_006234634.2 | XP_006234696.1 | 14 | 3883 | |||
| XM_006234635.2 | XP_006234697.1 | 14 | 2342 | |||
| XM_006234636.2 | XP_006234698.1 | 14 | 3333 | |||
| XM_008762060.1 | XP_008760282.1 | 13 | 2526 | |||
| GenBank mRNA | BC128741.1 | - | 12 | 2002 | 8363963 | |
| U69644.1 | - | 12 | 2222 | |||

Pax6
Genomic Map