siRNA Details
| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013005.1 | NP_037137.1 | 2, 3 | 435 | ||
| GenBank mRNA | D64045.1 | - | 2, 3 | 435 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pik3r1| Gene Symbol: Pik3r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013005.1 | NP_037137.1 | 2, 3 | 435 | ||
| GenBank mRNA | D64045.1 | - | 2, 3 | 435 | ||