siRNA Details
| Gene Symbol: Psap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190236.1 | NP_001177165.1 | 15 | 2100 | ||
| NM_001190237.1 | NP_001177166.1 | 15 | 2097 | |||
| NM_001190238.1 | NP_001177167.1 | 14 | 2088 | |||
| NM_013013.2 | NP_037145.2 | 14 | 2091 | |||
| XM_006256403.1 | XP_006256465.1 | 15 | 2109 | |||
| GenBank mRNA | BC061759.1 | - | 14 | 2022 | 5600088 | |
| FQ216707.1 | - | 3 | 583 | |||
| FQ217536.1 | - | 14 | 2046 | |||
| M19936.1 | - | 14 | 2040 | |||

Psap
Genomic Map