siRNA Details
| Gene Symbol: Sbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013024.2 | NP_037156.2 | 2 | 175 | ||
| XM_008767547.1 | XP_008765769.1 | 2 | 267 | |||
| GenBank mRNA | CA339008.1 | - | 2 | 175 | ||
| J02675.1 | - | 2 | 291 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sbp| Gene Symbol: Sbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013024.2 | NP_037156.2 | 2 | 175 | ||
| XM_008767547.1 | XP_008765769.1 | 2 | 267 | |||
| GenBank mRNA | CA339008.1 | - | 2 | 175 | ||
| J02675.1 | - | 2 | 291 | |||