siRNA Details
| Gene Symbol: Slc18a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013031.1 | NP_037163.1 | 3 | 537 | ||
| XM_006231664.2 | XP_006231726.1 | 3 | 1378 | |||
| XM_008760515.1 | XP_008758737.1 | 2 | 737 | |||
| XM_008760516.1 | XP_008758738.1 | 3 | 1383 | |||
| XM_008760517.1 | XP_008758739.1 | 3 | 1378 | |||
| XM_008760518.1 | XP_008758740.1 | 2 | 737 | |||
| XM_008760519.1 | XP_008758741.1 | 2 | 737 | |||
| XM_008760520.1 | XP_008758742.1 | 2 | 737 | |||
| GenBank mRNA | L00603.1 | - | 3 | 537 | ||
| M97381.1 | - | 3 | 508 | |||

Slc18a2
Genomic Map