siRNA Details
| Gene Symbol: Slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013033.2 | NP_037165.1 | 7 | 772 | ||
| GenBank mRNA | BC081827.1 | - | 7 | 804 | 7129017 | |
| D16101.1 | - | 7 | 772 | |||
| U03120.1 | - | 7 | 730 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc5a1| Gene Symbol: Slc5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013033.2 | NP_037165.1 | 7 | 772 | ||
| GenBank mRNA | BC081827.1 | - | 7 | 804 | 7129017 | |
| D16101.1 | - | 7 | 772 | |||
| U03120.1 | - | 7 | 730 | |||