siRNA Details
| Gene Symbol: Snai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013035.1 | NP_037167.1 | 2 | 162 | ||
| GenBank mRNA | AF497973.1 | - | 2 | 162 | ||
| U97061.1 | - | 1 | 96 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snai2| Gene Symbol: Snai2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013035.1 | NP_037167.1 | 2 | 162 | ||
| GenBank mRNA | AF497973.1 | - | 2 | 162 | ||
| U97061.1 | - | 1 | 96 | |||