siRNA Details
| Gene Symbol: Id3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013058.2 | NP_037190.1 | 3 | 531 | ||
| GenBank mRNA | BC064658.1 | - | 3 | 531 | 5623701 | |
| D10864.1 | - | 3 | 508 | |||
| FQ212997.1 | - | 3 | 553 | |||
| FQ216267.1 | - | 3 | 553 | |||
| FQ218468.1 | - | 3 | 551 | |||
| FQ218958.1 | - | 3 | 544 | |||
| FQ229332.1 | - | 3 | 553 | |||

Id3
Genomic Map