siRNA Details
| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013059.1 | NP_037191.1 | 12 | 1997 | ||
| XM_006239136.2 | XP_006239198.1 | 12 | 2371 | |||
| GenBank mRNA | BC088399.1 | - | 12 | 2020 | 7128677 | |
| J03572.1 | - | 12 | 1997 | |||
| Y00714.1 | - | 11 | 1841 | |||

Alpl
Genomic Map