siRNA Details
| Gene Symbol: Ppp1cb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013065.2 | NP_037197.1 | 2 | 366 | ||
| XM_008764505.1 | XP_008762727.1 | 3 | 304 | |||
| GenBank mRNA | BC062033.1 | - | 2 | 366 | 5623630 | |
| D90164.1 | - | 2 | 146 | |||
| S78218.1 | - | 2 | 133 | |||

Ppp1cb
Genomic Map