siRNA Details
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB100593.1 | - | 3 | 337 | ||
AB100594.1 | - | 3 | 337 | |||
BC061756.1 | - | 3 | 341 | 5600029 | ||
X05300.1 | - | 3 | 342 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB100593.1 | - | 3 | 337 | ||
AB100594.1 | - | 3 | 337 | |||
BC061756.1 | - | 3 | 341 | 5600029 | ||
X05300.1 | - | 3 | 342 |