siRNA Details
| Gene Symbol: Lep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013076.3 | NP_037208.1 | 1 | 154 | ||
| XM_008762762.1 | XP_008760984.1 | 1 | 242 | |||
| GenBank mRNA | D45862.1 | - | 2 | 166 | ||
| D49653.1 | - | 1 | 135 | |||
| S78586.1 | - | 1 | 106 | |||
| U48849.1 | - | 1 | 67 | |||

Lep
Genomic Map