siRNA Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013084.2 | NP_037216.1 | 2 | 143 | ||
| XM_008759865.1 | XP_008758087.1 | 2 | 228 | |||
| XM_008759866.1 | XP_008758088.1 | 2 | 228 | |||
| XM_008759867.1 | XP_008758089.1 | 2 | 229 | |||
| GenBank mRNA | U64451.1 | - | 2 | 144 | ||

Acadsb
Genomic Map