siRNA Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013084.2 | NP_037216.1 | 3 | 244 | ||
| XM_008759865.1 | XP_008758087.1 | 3 | 329 | |||
| XM_008759866.1 | XP_008758088.1 | 3 | 329 | |||
| XM_008759867.1 | XP_008758089.1 | 3 | 330 | |||
| GenBank mRNA | FQ210776.1 | - | 3 | 253 | ||
| U64451.1 | - | 3 | 245 | |||

Acadsb
Genomic Map